VeriSpec™ Gallium (Ga) Standard for AAS 1000 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010020-100N RV010020-500N
RCRV010020100NEA 282.82 CAD
RCRV010020100N RCRV010020500N
VeriSpec™ Gallium (Ga) Standard for AAS 1000 ppm in 2% HNO₃
Standards Cationic Standards Gallium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility