VeriSpec™ Erbium (Er) Standard for AAS 1000 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010017-100N RV010017-500N
RCRV010017100NEA 158 CAD
RCRV010017100N RCRV010017500N
VeriSpec™ Erbium (Er) Standard for AAS 1000 ppm in 2% HNO₃
Standards Cationic Standards Erbium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility