VeriSpec™ Dysprosium (Dy) Standard for AAS 1000 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010016-100N
RV010016-500N
RCRV010016100NEA
158
CAD
RCRV010016100N
RCRV010016500N
VeriSpec™ Dysprosium (Dy) Standard for AAS 1000 ppm in 2% HNO₃
Standards
Cationic Standards
Dysprosium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility