VeriSpec® Copper (Cu) Standard for AAS 1000 ppm in 2% HCl, RICCA Chemical Company
Supplier: Ricca Chemical
RV010015-100N RV010015-500N
RCRV010015-1NEA 228.06 CAD
RCRV010015-1N RCRV010015100N RCRV010015500N
VeriSpec® Copper (Cu) Standard for AAS 1000 ppm in 2% HCl, RICCA Chemical Company
Standards Cationic Standards Copper Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility