VeriSpec® Cesium (Cs) Standard for AAS 1000 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010014-100N RV010014-500N
RCRV010014100NEA 126.4 CAD
RCRV010014100N RCRV010014500N
VeriSpec® Cesium (Cs) Standard for AAS 1000 ppm in 2% HNO₃
Standards Cationic Standards Cesium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility