VeriSpec® Cobalt (Co) Standard for AAS 1000 ppm in 2% HCl
Supplier: Ricca Chemical
RV010012-100N RV010012-500N
RCRV010012-1NEA 244.24 CAD
RCRV010012-1N RCRV010012100N RCRV010012500N
VeriSpec® Cobalt (Co) Standard for AAS 1000 ppm in 2% HCl
Standards Cationic Standards Cobalt Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility