VeriSpec® Calcium (Ca) Standard for AAS 1000 ppm in 2% HCl, RICCA Chemical Company
Supplier: Ricca Chemical
RV010009-100N RV010009-500N
RCRV010009-1NEA 247 CAD
RCRV010009-1N RCRV010009100N RCRV010009500N
VeriSpec® Calcium (Ca) Standard for AAS 1000 ppm in 2% HCl, RICCA Chemical Company
Standards Cationic Standards Calcium Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility