VeriSpec™ Gold (Au) Standard for AAS 1000 ppm in 2% HCl
Supplier: Ricca Chemical
RV010004-100C RV010004-500C
RCRV010004100CEA 426.6 CAD
RCRV010004100C RCRV010004500C
VeriSpec™ Gold (Au) Standard for AAS 1000 ppm in 2% HCl
Standards Cationic Standards Gold Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility