VeriSpec® Silver (Ag) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Supplier: Ricca Chemical
RV010001-100N RV010001-500N
RCRV010001100NEA 120.08 CAD
RCRV010001100N RCRV010001500N
VeriSpec® Silver (Ag) Standard for AAS 1000 ppm in 2% HNO₃, RICCA Chemical Company
Standards Cationic Standards Silver Standards
Certified reference material (CRM) for AA (atomic absorption)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility