SP Wilmad-LabGlass Hempel Distilling Columns with Vacuum Jacket, SP Industries
Supplier: SP Industries
LG-5792-124 LG-5792-140
14200-660EA 2172.45 CAD
14200-660 14200-980
SP Wilmad-LabGlass Hempel Distilling Columns with Vacuum Jacket, SP Industries
Distillation Components Distillation Columns
These Hempel Columns are enclosed in an evacuated jacket which is completely silvered except for two narrow clear strips 180° apart that serve as windows.
  • Made of borosilicate glass
The column consists of a plain tube with full length Standard Taper inner joint at bottom and Standard Taper outer joint of the same size at top. Near the bottom there is a row of indentations to support random type packing material.