ECO Funnels, Chemglass
Supplier: Chemglass
CG-1759-02 CG-1759-03 CG-1759-04 CG-1759-06 CG-1759-26 CG-1759-01
33501-494EA 534.59 CAD
33501-494 33501-496 33501-498 33501-500 33501-518 80061-538
ECO Funnels, Chemglass
Funnels Filtration Funnels Filter Funnels
Eco Funnels are made of HDPE.
Funnels feature an easy to use lid. A gasket and positive latch close the lid and maintain an airtight seal. A filter is also built into the interior to reduce the loss of magnetic stir bars and other items when emptying the contents of a vessel into the funnel. The funnel is equipped with an 11" stem to reduce chemical evaporation of hazardous solvents in the lab.