Magnesium nitrate solution 20 g/L in water, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy

Supplier: Ricca Chemical
Danger

RV010136-50N
RCRV010136-50NEA 657.22 CAD
RCRV010136-50N
Magnesium nitrate solution 20 g/L in water, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy
Magnesium nitrate solutions

Certifications: Manufactured and Tested in an ISO 17025/Guide 34 Accredited Facility

Formula: Mg(NO₃)₂
CAS Number: 10377-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR