Magnesium nitrate solution 10000 ppm in water, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy

Supplier: Ricca Chemical
Danger

RV010130-50N RV010130-100N
RCRV010130-50NEA 453.84 CAD
RCRV010130-50N RCRV010130100N
Magnesium nitrate solution 10000 ppm in water, VeriSpec™ matrix modifier solution, for atomic absorption spectroscopy
Magnesium nitrate solutions
Formula: Mg(NO₃)₂
Storage Temperature: Ambient
CAS Number: 10377-60-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR