NORELL™ NMR Tubes, Valved, Chemglass
Supplier: Chemglass
C-S-5-600-JY-7 C-S-5-600-JY-8 C-S-5-500-JY-7 C-S-5-500-JY-8
16004-740EA 547.04 CAD
16004-740 16004-742 16004-744 16004-746 16004-748 16004-750 89209-448 89209-450
NORELL™ NMR Tubes, Valved, Chemglass
Tubes NMR Tubes
NORELL™ Valved NMR Tubes are a completely greaseless PTFE assembly, which is easy to use and to disassemble for cleaning.
  • PTFE covered O-ring eliminates material incompatibilities
  • Can be handled without flame-sealing tubes
  • Includes female joint for quick attachment to vacuum rack