White Polypropylene Screw Caps, F217 & Fluoropolymer Resin Liner, Qorpak®
Supplier: Qorpak
239081 239082 239084 239085 239087 272188 272190 272191 272192 272193 272194 272195 272196 272197 272198 272199 272200 272201 272202 272203 272204 239077 239080 239083 239086 239071 239075 239088 239089 239090 262535 239355
75799-974CS 2200.69 CAD
75799-974 75799-976 75799-978 75799-980 75799-982 76012-826 76012-830 76012-832 76012-834 76012-836 76012-838 76012-840 76012-842 76012-844 76012-846 76012-848 76012-850 76012-852 76012-854 76012-856 76012-858 76170-280 76170-282 76170-284 76170-286 76170-300 76170-302 76170-304 76170-306 76170-308 76170-390 76170-392 76170-412
White Polypropylene Screw Caps, F217 & Fluoropolymer Resin Liner, Qorpak®
Closures Bottle Closures
Polypropylene caps with F217 & Fluoropolymer Resin liners are an economical alternative to Thermoset & Phenolic Fluoropolymer Resin lined caps.
  • Liners are very resilient – great sealing capability
  • Liners have strong resistance to chemicals and moisture
Some uses for these F-217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.