Prosolv® Recycling System and Components, Justrite®
Supplier: Justrite
28191 28190
75870-998EA 477.24 CAD
75870-998 75870-996
Prosolv® Recycling System and Components, Justrite®
Safety Cans
Safely and economically convert gas cylinders into recyclable steel containers.
  • Prosolv® System removes the sealing sleeve and internal valve stem from propane, MAPP, and 0.75" male threaded calibration gas cylinders
  • Empties and filters propellants from cylinders, converting them into non-hazardous recyclable material
  • Not for use with oxygen bottles
Spent propane canisters are considered hazardous waste while still under pressure, but once they are safely vented and relieved of pressure they can be recycled as scrap steel. The Prosolv® System safely recycles depressurized propane cylinders, eliminating an entire category of hazardous waste from the waste stream.
Certifications: EPA Compliant.
Accessories information: System includes vent device, carbon filter, antistatic wire, safety goggles, sleeve removal tool, and 40 certification tags.