ASTM Class 1 Weight Sets, METTLER TOLEDO®
Supplier: Mettler Toledo
11124002 11124004 11124006 11124008 11124014 11124018 11124001 11124003 11124005 11124007 11124009 11124015 11124019 11124000
CA97021-840EA 2305.18 CAD
CA97021-840 CA97021-842 CA97021-844 CA97021-846 CA97021-848 CA97021-850 CA97021-852 CA97021-854 CA97021-856 CA97021-858 CA97021-860 CA97021-862 CA97021-864 97021-838
ASTM Class 1 Weight Sets, METTLER TOLEDO®
Weights Calibration Weights
These ASTM weight sets with adjusting cavity are made of vaccum-melted stainless steel, with an electropolished finish.
  • Electrolytic polishing ensures glossy surfaces for anti-adhesion effects
  • Exclusive material has a density of 7.9 kg/dm3