Weighing/Transfer Funnels, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
747585-0013 747585-0020
KT747585-0013EA 136.4 CAD
KT747585-0013 KT747585-0020
Weighing/Transfer Funnels, Kimble Chase, DWK Life Sciences
Funnels Filtration Funnels Filter Funnels
These funnels are designed for use with micro volumetric flasks and are ideal for the easy transfer of liquids or powders.
  • Flat bottom allows weighing
  • Manufactured from 33 expansion low extractable borosilicate glass
Accessory for microscale MICROFLEX® threaded kits.
Certifications: Conforms to USP Type I and ASTM E438, Type I, Class A requirements.