xtra ST Industrial Ultrasonic Cleaners, Elma
Supplier: Elma
1077060 1077481 1077482 1077483 1077056 1077058 1077059
75848-764EA 49606.11 CAD
75848-764 75848-810 75848-812 75848-814 75848-816 75848-818 75848-820
xtra ST Industrial Ultrasonic Cleaners, Elma
Ultrasonic Cleaners
Rugged Elmasonic xtra ST high capacity ultrasonic cleaners are designed for cleaning a wide range of parts in a production environment.
  • Powerful heavy-duty ultrasonic cleaner
  • Portable and easy to use
  • Dual frequency for coarse and fine cleaning
  • Sweep mode evenly distributes cleaning
  • Pulse mode adds bursts of ultrasonic power for intensive cleaning and degassing
  • Dynamic mode combines Sweep and Pulse for optimum cleaning results
Stubborn contaminants like polishing pastes are easily removed at 25 kHz
The stainless steel tanks are wheel mounted forMore Product Information