E Plus Ultrasonic Bath with Pulse Mode for Mixing, Cleaning, and Degassing, Elma
Supplier: Elma
1071665 1071667 1071657 1071649 1071670 1071647 1071676 1071672 1071674 1071653
75848-752EA 3749.34 CAD
75848-752 75848-740 75848-754 75848-742 75848-750 75848-748 75848-756 75848-744 75848-758 75848-746
E Plus Ultrasonic Bath with Pulse Mode for Mixing, Cleaning, and Degassing, Elma
Ultrasonic Cleaners
The 37 kHz Elmasonic E Plus series includes 9 tank sizes with simple, user-friendly operation. Samples are rapidly mixed, dispersed, dissolved, or homogenized in Pulse mode.
  • Thorough and uniform cleaning with Sweep function
  • Unique Pulse function performs laboratory mixing and degassing operations
  • Pulse function also provides powerful cleaning for difficult jobs
  • Quality promise: 2-year warranty with replacment guarantee for the first year
Pulse mode boosts the ultrasonic power to remove tenacious contaminants such as mineral deposits and polishing pastes. Sweep mode produces a continuous slight variation of frequency to provide uniform and thorough cleaning of parts.
The microprocessor-controlled generator in this sonic cleaner maintains constant ultrasonic power, sensing and adjusting for light or heavy loads in the tank. Digital operation with power tracking ensures reproducible performance from run to run.
All models (except the EP10) have adjustable bath temperature from 30°C to 80°C. The control panel indicates when the set temperature has been reached. The timer can be set from 1-30 minutes or for continuous operation with manual shut off. The unit turns off automatically after 8 hours of continuous operation for safety. The 1.5 gallon unit and upwards have a rear drain with a valve on the side of the tank for easy access. The tank cover can be flipped over and used as a drip tray.
Dependable, state-of-the-art Elma ultrasonic cleaners are made in Germany.
Certifications: UL, CSA, CE, ISO9001, ISO13485
Delivery information: All models are supplied with power cord, plastic lid and manual. Baskets must be ordered separately. Optional stainless steel baskets have plastic coated handles for insulation and noise reduction.