CONTEXT™ Cleanroom Documentation Paper and Notebooks, Contec®
Supplier: Contec
CNBK5585W CNTX1117B CNTX1117W CNTX8511B CNBK8511W CNTXA4B CNTX8511G CNTX8511P CNTX8511W CNTXA4W CNTX8511Y
75846-270CS 366.09 CAD
75846-270 75846-272 75846-274 75846-276 75846-278 75846-280 75846-282 75846-284 75846-286 75846-288 75846-290
CONTEXT™ Cleanroom Documentation Paper and Notebooks, Contec®
Paper
CONTEXT™ Cleanroom Documentation offers a wide range of stationery products that reduce the risk of particle and fiber release. Designed for use in controlled environments, CONTEXT™ uses Munising® Bond material made from synthetic polymer-saturated cellulose to provide optimal smoothness and naturally enhanced writeability. Free of inorganic fillers and latex, it poses no risk to operators.
  • Synthetic polymer-saturated cellulose reduces the number of particles released during use
  • No inorganic fillers or natural latex, preventing latex reactions
  • Heat
More Product Information