Round Metal Specimen Chucks, Suitable for Shandon Cryostats, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
SHD30YL SHD30WT SHD30PK SHD30GN SHD30OR SHD40RD SHD40BK SHD40BL SHD40YL SHD40WT SHD40PK SHD40GN SHD40OR SHD30RD SHD30BK SHD30BL
75846-780EA 184.51 CAD
75846-780 75846-782 75846-784 75846-786 75846-788 75846-790 75846-792 75846-794 75846-796 75846-798 75846-800 75846-802 75846-804 75846-836 75846-838 75846-840
Round Metal Specimen Chucks, Suitable for Shandon Cryostats, Springside Scientific
Specimen Mounting Supplies
Round metal specimen chuck with O-ring made of low temperature resistant rubble; with stem length ~37.5 mm.
  • Different o-ring colors eliminates sample errors and the use of expensive colored freezing medium
  • Orientation mark to assure proper sample positioning
Proper dimensional tolerance and soft aluminum, to avoid slippage slip in chuck that results in thin/thick cutting.