Flat Round Metal Specimen Chucks, Suitable for Thermo Cryostars, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
CRSTAR30RD CRSTAR30BK CRSTAR30BL CRSTAR30YL CRSTAR30WT CRSTAR30PK CRSTAR30GN CRSTAR30OR
75846-820EA 203.16 CAD
75846-820 75846-822 75846-824 75846-826 75846-828 75846-830 75846-832 75846-834
Flat Round Metal Specimen Chucks, Suitable for Thermo Cryostars, Springside Scientific
Seals Sealing Rings
Flat round metal specimen chuck O-ring made of low temperature resistant rubble with 30 mm work area.
  • Different o-ring colors eliminates sample errors and the use of expensive colored freezing medium
  • Orientation mark to assure proper sample positioning
Proper dimensional tolerance and soft aluminum, to avoid slippage slip in chuck that results in thin/thick cutting.