TempDex 710, Nitrile Gloves, Mapa Professional
Supplier: MAPA
34710037 34710039
10067-640CS 704.7 CAD
10067-640 10067-642 10067-644
TempDex 710, Nitrile Gloves, Mapa Professional
Gloves
These high dexterity and thermal protection gloves are suitable for handling of composite materials after heat processing in ceramics, plastics and composite industries.
  • Nitrile coated palm and dotted embossing for better insulation and grip
  • Thin, anatomically-shaped glove ensures dexterity
  • Long lasting and abrasion resistant
Ideal for use in ceramics, plastics, composite and construction industries.