Hydranal™ Water Standards for Karl Fischer Titration, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
34696-6X25G 34828-6X40ML 34847-6X40ML 34849-6X80ML 34446-40ML 34748-6X10G 34813-6X100ML 34813-6X500ML
BJ34696-25GCS 404.53 CAD
BJ34696-25G BJ34828-40ML BJ34847-40ML BJ34849-80ML BJ34446-40ML BJ34748-6X10G BJ34813-6100ML BJ34813-6500ML
Hydranal™ Water Standards for Karl Fischer Titration, Honeywell Research Chemicals
Standards Water Determination (by Karl Fischer method) Standards
Broad product range for volumetric and coulometric Karl Fischer applications, as well as the indirect oven application.
  • Manufactured according to ISO 9001 requirements
  • Report of Analysis showing the exact water content and expanded uncertainty
  • Verified against external standards such as NIST SRM 2890 or NMIJ CRM 4222
  • Long shelf life (up to five years)
  • Convenient packaging
Delivery information: Supplied with detailed instruction for use.
Caution: The recommended storage temperature for product HYDRANAL™ Water Standard is 2-8°C. The product can be transported at normal ambient temperatures. In the long term, temperatures > 20°C (if possible > 8 °C) should be avoided.