Orion™ Powder Chemistry Packs, Thermo Scientific
Supplier: Thermo Fisher Scientific
Orion™ AQUAfast Powder Pack Reagents perform colorimetric measurements of common water parameters with powders that dissolve quickly and mix easily.
- Equivalent to other widely used and recognized chemistries
- Compatible with Orion™ AQUAfast Colorimeters
Orion™ AQUAfast Powder Pack Reagents are designed for use with Orion™ AQUAfast colorimeters and Orion™ AquaMate Vis and UV-Vis spectrophotometers.
Powder pack chemistries dissolve quickly in solution, are easy to mix, and do not require a stir rod or crushing. Each powder pack is notched for simplified opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica, and sulfate.