Orion™ ROSS Ultra™ Glass Triode™ pH/ATC Combination Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
8302BNUMD 8302BNURCA 8302BNURCA
89187-024EA 1224.65 CAD
89187-024 10324-314 CA10324-314
Orion™ ROSS Ultra™ Glass Triode™ pH/ATC Combination Electrodes, Thermo Scientific
Electrodes pH Electrodes
No more waiting for pH readings to stabilize when measuring samples of varying temperatures.
  • Measure pH and temperature with one electrode
  • pH range of 0 to 14
  • Built-in temperature sensors offer a convenient solution for routine sample measurements
  • For use in Pharmaceutical, Food Manufacturing, Academic/Government, and Environmental industries
Orion™ ROSS Ultra™ Triode™pH electrodes combine the stability, precision, and speed of response of the ROSS Ultra™ line with integrated temperature measurement. These electrodes contain no Silver or Silver Chloride and incorporate a double-junction design, which protects the sample from contamination and makes the electrodes compatible with TRIS buffers.