Orion™ pH Buffer Cubitainers™, Thermo Scientific
Supplier: Thermo Fisher Scientific
9110CB
9107CB
9104CB
23197-994EA
276.8
CAD
23197-994
23197-996
23197-998
Orion™ pH Buffer Cubitainers™, Thermo Scientific
Buffers
pH Calibration Buffers
Hold 5 gallons (19L) of pH buffer in color-coded Orion™ pH Buffer Cubitainers™
- Designed for for accurate and reproducible pH calibrations
- Color coded to simplify identification and selection
- Packaged with buffer/temperature tables for accurate calibrations and NIST traceability
- Manufactured under ISO 9000 quality standards
Each vessel is equipped with an easy-pour spigot.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...