Ice Borer, Bürkle
Supplier: BURKLE INC
5323-0200 5323-2010 5323-0190
75806-594EA 1183.03 CAD
75806-594 75806-596 76501-442
Ice Borer, Bürkle
Samplers Solid Samplers
Sampler for deep-frozen and semi-solid substances products in the food industry.
  • Stainless steel AISI 316L (1.4404) / AISI 316 Ti (1.4571)
  • Sterilizable
  • Complete with case
The sharp sawing crown of the hollow borer rapidly cuts into the sample
It consists of three parts: borer, borer head for use in conventional drills and rod for pressing the cylindrical sample out of the borer head.
A cylindrical sample (cross sectional sample) with a diameter of 1.6 cm and a length of up to 20 cm can be cut out.