Orion™ AQUAfast™ IV Chemistry Reagents, Thermo Scientific
Supplier: Thermo Fisher Scientific
AC4004 AC4005 AC4007 AC4095 AC4016 AC4012 AC4099 AC4006 AC4078 AC4065 AC4070 AC4046 AC4048 AC4055 AC4011 AC4082
CA14217-844PK 575.26 CAD
CA14217-844 CA14217-846 CA14217-848 CA14217-850 CA14217-856 CA14217-858 CA14217-864 CA14217-866 CA14217-868 CA14217-870 CA14217-876 CA14217-878 CA14232-930 CA14232-932 CA14232-934 CA14232-938 14232-937
Orion™ AQUAfast™ IV Chemistry Reagents, Thermo Scientific
Environmental Test Kits
Orion™ AQUAfast™ Auto-Test Ampoule Reagents can be used with the Orion™ AQUAfast™ AQ4000 colorimeter to simplify sample preparation and tests.
  • Consistency and reproducibility improved with AQUAfast™ ampoules and an AQ4000 meter
  • Auto-ID™ eliminates measurement errors
  • Designed for use with colorimetric water and wastewater testing equipment
The Auto-Test ampoules contain a pre-measured amount of liquid reagent and easily fill with the correct volume of sample for simple and fast sample and reagent mixing. Each ampoule contains an Auto-ID barcode that is read by the AQ4000 colorimeter and used to automatically identify the test parameter and initiates the test.