SP Wilmad-LabGlass 90° Bend Adapters with Hose Connection and Standard Taper Joint, SP Industries
Supplier: SP Industries
ML-195-700
14202-808EA 78.67 CAD
14202-808
SP Wilmad-LabGlass 90° Bend Adapters with Hose Connection and Standard Taper Joint, SP Industries
Adapters Adapters, Joints with Tube Connectors
Featuring a 90° bend to hose connector at top and a Standard Taper inner joint at bottom.
  • 90° angle bend
  • Top hose connector
  • Bottom inner [ST] joint
Adapters are made of borosilicate glass.