VWR® Traceable® Memory-Loc™ USB Datalogging ULT Freezer Thermometer
Supplier: VWR International
75784-092 75784-094 75999-320 75999-322 75999-340
75784-092EA 769.32 CAD
75784-092 75784-094 75999-320 75999-322 75999-340
VWR® Traceable® Memory-Loc™ USB Datalogging ULT Freezer Thermometer
Data Loggers
USB Datalogging ULT freezer thermometer that can be used in 21 CFR 11 environments raw data is locked and may not be cleared/changed on base unit.
  • Meets current CDC requirements for vaccine storage and monitoring
  • Rolling memory structure, maintains most recent one million (1,048,576) temperature observations
  • Smart-Alarm™ features a visual/audible alarm that continues to alarm even if the unit returns to non-alarm conditions
  • Stores up to ten unique alarm events
  • Memory can be cleared on unit after downloading data to USB stick
  • Hassle-free retrieval of data -
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Interface USB USB
Weight 142 g 142 g 142 g 142 g
W×D×H 108×70×19 mm (4.25×2.75×0.75") 108×70×19 mm (4.25×2.75×0.75")