Histo Plas™ Embedding Rings, Bio Plas, Inc
Supplier: Bio Plas
6003 6002 6001
21150-002CS 1678.56 CAD
21150-002 21150-006 21150-004
Histo Plas™ Embedding Rings, Bio Plas, Inc
Embedding Cassettes Embedding Rings
Histology Embedding Rings provide a quick and easy way to uniformly embed tissues for further processing.
  • Uniform Block Size For Sectioning
  • Work With Stainless Steel Or Plastic Base Molds
  • Provide Convenient Storage And Identification
Simply place the embedding ring atop a base mold, fill with paraffin, and allow to cool
Top quality, made in the USA.
Available in Three colors for color coding references.