Dipping vessel, Burkle
Supplier: BURKLE INC
5365-1000 5365-2000
75846-228EA 2046.59 CAD
75846-228 75846-296
Dipping vessel, Burkle
Samplers Fluid Samplers
Dipping vessel for taking all-layer samples from tanks, tank cars, water courses, water purification plants.
  • Equipped with Easy-Flow; a flow optimized flow valve
  • High weight (2.1 kg/4.63 lbs.) for faster lowering
  • Height with handle is 427 mm (16.81")
  • Made of non-sparking brass or stainless steel AISI 304 (1.4301) for Ex area
  • Tested for use in the Ex Protection Zone 0
  • Explosion Categories IIA, IIB, and IIC
Packaging: Supplied one dipping vessel per package.