Metzenbaum-Lahey Dissecting Scissors, OR Grade, Current Censitrac Customers Only, Sklar
Supplier: Sklar
22-1057 22-1070 22-1071
89025-202EA 272.76 CAD
89025-202 89025-206 89025-208
Metzenbaum-Lahey Dissecting Scissors, OR Grade, Current Censitrac Customers Only, Sklar
Scissors
Metzenbaum-Lahey Dissecting Scissors are most commonly used for dissecting and cutting tissue.
These scissors are not recommended for cutting sutures, drains, or heavy tissue
Censitrac Ready OR Grade Metzenbaum-Lahey Dissecting Scissors are industry standard and constructed of stainless steel.
Metzenbaum-Lahey scissors tend to be shorter than traditional Metzenbaum scissors.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.