BLAUBRAND® Graduated Pipettes, Class AS, USP, Certified, BrandTech
Supplier: Brandtech
27506 27509 27511 27513
10218-684PK 229.5 CAD
10218-684 10218-686 10218-688 10218-690
BLAUBRAND® Graduated Pipettes, Class AS, USP, Certified, BrandTech
Pipettes Graduated Pipettes
Volumetrically stable even when heated up to 250 °C via special annealing process.
  • Certified Class A
  • Calibrated to deliver (TD, Ex.)
  • Constructed according to DIN EN ISO standards with error limits that meet the specifications of USP/ASTM
  • Flexible screen stenciled graduations for maximal accuracy at intermediate volumes
  • Durable, high-contrast blue enamel markings
Each individual piece is marked on a computer controlled, automated production line to ensure maximum precision.
Certifications: Class AS per DIN EN ISO 835. Meets standards of USP Section 3More Product Information