Silylation Reagents
Supplier: Thermo Scientific
TS-38834 TS-38831 TS-38832 TS-38833 TS-38840
10056-196EA 889.48 CAD
10056-196 10066-314 10066-316 10066-318 10066-322
Silylation Reagents
Chromatography Sorbents GC Sorbents
BSTFA + 1% TMCS is ideal for derivatizing fatty acid amides, slightly hindered hydroxyls and other difficult-to silylate compounds.
  • Derivate fatty acid amides
  • Catalyzed formulation
  • Stronger than BSTFA
This catalyzed formulation is stronger than BSTFA alone.
Storage temperature: 2 to 8 °C