Ultrapure Straight Plug Stoppers, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
W224100-400 W224100-405
10796-552CS 586.11 CAD
10796-552 10796-556
Ultrapure Straight Plug Stoppers, Wheaton
Stoppers Rubber Stoppers
Manufactured from an ultrapure bromobutyl formulation with extremely high chemical purity and low gas permeability.
  • Low particulate content and low level of extractables
  • Free of latex, nitrosamines, and 2-MCBT (2- mercaptobenzothiazole)
  • Ideal for "ready for sterilization" and "ready for use" applications
  • Autoclavable
Certifications: Complies with US, European, and Japanese Pharmacopeias.