Support Extension, Glas-Col®, LLC
Supplier: Glas-Col
100EO545 100EO540 100EO550
33787-630EA 367.51 CAD
33787-630 33793-092 33793-102
Support Extension, Glas-Col®, LLC
Retort Stands
Table top support for rounded bottom fabric mantles.
  • Steel support ring
  • Fabric strap basket cradle
  • Durable construction
  • For use with round bottom soft shell mantle
Our table top support holds spherical and hemispherical mantles up to 5 L. Attaches easily to ring stands or distillation racks.
Delivery information: 7 to 10 days after order.
Order Now
SPECIFICATIONS
Environmentally Preferable