Heating Tape, Tubes and Pipes, Glas-Col®
Supplier: Glas-Col
103ADETO.56 103ADETO.510 103ADETO.54 103ADETO.58 103ADET120
33729-043EA 356.31 CAD
33729-043 33729-087 33729-021 33729-065 33730-268
Heating Tape, Tubes and Pipes, Glas-Col®
Tapes
Heat tape wrap for glass tubes and metal pipes in lengths 4 to 20 foot.
  • Ideal for use in high moisture areas
  • Resisitant to most laboratory environmental conditions
  • Passes 1,000V electrical insulation test
Our heating tapes are designed to heat odd lengths of glass tubes and metal pipes. With ribbon type heating elements running lengthwise in the tape and encased in durable glass braid coated in silicone rubber they are ideal for use in high moisture areas. Tapes are sealed at both ends with waterproof fittings. Temperature limit of 480 degrees F.
Delivery information: 7-10 days after order.