Heating Mantles, High Temp, Glas-Col®
Supplier: Glas-Col
100BSTM1000 100B STM1100 100B STM900
33752-450EA 1372.56 CAD
33752-450 89260-002 89260-004
Heating Mantles, High Temp, Glas-Col®
Heaters Heating Mantles
Aluminum housed mantle for applications requiring more heat.
  • Fabric interior
  • Durable aluminum outer shell
  • Adaptable to most vessels
The Series "STM" mantle is designed for applications requiring a higher temperature. The interior is resilient glass fabric which nesltles the glassware. The exterior is rigid aluminum providing strength and support. The mantle has an internal operating temp of up to 600 degrees C.
Delivery information: 7-10 days after order.