Insulating Mantle Top, Glas-Col®
Supplier: Glas-Col
100AINS1103 100AINS1143 100AINS1083 100AINS1063 100AINS1123 100AINS1163
33787-514EA 1042.74 CAD
33787-514 33787-516 33787-506 89260-030 33787-510 33787-512
Insulating Mantle Top, Glas-Col®
Heaters Heating Mantles
Insulate and help prevent vapor condensation.
  • Protects flask during process
  • Laced side opening
  • Elliptical hole accomodates 1, 2, and 3 neck flasks
This mantle does not heat but insulates and helps prevent vapor condensation in the upper half of a flask during distillation of high boiling materials. Our insulating tops are Series M, TM, or STM heating mantles. They protect the flask, the solutions and the laboratory.
Ordering information: Insulating top only.
Delivery information: 7-10 days after order.