Heating Mantle Top, Top Only, Glas-Col®
Supplier: Glas-Col
100AMO1083 100AMO1123 100AMO1063 100AMO1143 100AMO1163
33779-295EA 934.85 CAD
33779-295 33779-389 33779-289 33779-369 33779-380
Heating Mantle Top, Top Only, Glas-Col®
Heaters Heating Mantles
Prevents vapor condensation in top half of flask during distillation.
  • Protects flask during process
  • Laced side opening
  • Elliptical hole accomodates 1, 2 and 3 neck flasks
Our heated top mantle covers the upper half of a flask during distillation of high boiling materials. The tops are designed for use with our M, TM or STM mantles. They protect the flask from adverse conditions which would disturb the reflux ration during fractionation.
Delivery information: 7-10 days after order.