Stir Mantles, Glas-Col®
Supplier: Glas-Col
100DTEM102 100DTEM104 100DTEM106 100DTEM108 100DTEM112
33787-302EA 1814.84 CAD
33787-302 33787-304 33787-306 33787-308 33787-312
Stir Mantles, Glas-Col®
Heaters Heating Mantles
Heating and stirring in one unit.
  • Electromagnetic
  • Independent heating and stirring
  • Rigid housing for strength and support
  • For use with spherical flask
This system has the power to mix small volumes in a flask without an overhead stirrer. Electromagnetic speed range of 50-750 rpm. Mantle section is aluminum housed with fabric interior liner to softly nestle glassware. Ideal for temperatures up to 400 degrees C.
Delivery information: 7-10 days after order.