FreeZone® Stoppering and Bulk Tray Dryer Accessories, Labconco®
Supplier: Labconco
7204300 7377900 76000-960 76000-962 76000-964 76000-966
75840-986EA 11297 CAD
75840-986 75841-024 76000-960 76000-962 76000-964 76000-966
FreeZone® Stoppering and Bulk Tray Dryer Accessories, Labconco®
Freeze Dryers
Various accessories for FreeZone® stoppering and bulk tray dryers.
The small tray makes sample loading and unloading as well was cleaning easy when using the FreeZone® Small Tray Dryer (sold separately).
The rectangular test tube rack holds up to (70) 12 to 13 mm tubes for freeze drying. Made of type 304 stainless steel and includes bottom. It's ideal for use with the FreeZone® Small Tray Dryer, Bulk Tray Dryer or Stoppering Tray Dryer.
The rectangular test tube/microcentrifuge rack also holds up to (70) 12 to 13 mm tubes. It includes the rectangular test tube rack as well as a bottom and an adapter plate for 70 microcentrifuge tubes. Made of type 304 stainless steel. It is ideal for use with the FreeZone® Small Tray Dryer, Bulk Tray Dryer or Stoppering Tray Dryer.
Stainless steel shelves include 200-watt heater and sensor probe. FreeZone Bulk and Small Tray Dryers include three and can accommodate two additional Heated Shelves with Sensors. The Sensor, included with the Heated Shelf, may be used to monitor sample or shelf temperature.
Ordering information: Glassware, Tray Dryer/Stoppering Tray Dryer, and Freeze Dryer are required (not included).