Roc LC Guard Column Cartridges, Restek
Supplier: Restek
953550210 953050210 953650210 953450210 953350210
75846-082PK 311.18 CAD
75846-082 75846-104 75846-126 75846-148 75846-170
Roc LC Guard Column Cartridges, Restek
Chromatography Columns
Protect your Roc LC columns with minimal effect on retention, peak shape, or efficiency.
Guard column cartridges require Roc guard column holder.