Roc Cyano HPLC Columns (USP L10), Restek

Supplier: Restek
9536315 9536365 9536515 9536565 9536575 953631E 953636E 953651E 953656E 953657E
75846-116EA 851.45 CAD
75846-116 75846-118 75846-120 75846-122 75846-124 75846-128 75846-130 75846-132 75846-134 75846-136
Roc Cyano HPLC Columns (USP L10), Restek
Chromatography Columns
The Restek Cyano is a traditional monomeric cyanopropylsilane that is recommended for assays where alternate selectivity, or confirmation, to a C18 or C8 column is desired.

  • USP phase code: L10
  • Ligand type: cyanopropyl
  • Surface area: 300 m²/g
  • Carbon load: 8%
  • End-cap: yes
  • pH range: 2.5 to 8.0
  • Maximum temperature: 80 °C

Roc HPLC columns are built to be the cornerstone for your LC lab. These solid, reliable columns are pressure rated for any 400 bar HPLC system and deliver the peak shape, reproducibility, ruggedness, and performance you demand for all of your conventional HPLC applications. An exceptional value, Roc columns are priced for bulk use in routine analyses and large-volume workflows.

It can be used in normal-phase, reversed-phase (best with mobile phase pH between 5 and 7), and HILIC modes. It is an excellent choice for the analysis of protonated bases.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR