VITLAB® VITgrip™ Laboratory Bottles, Polypropylene, BrandTech
Supplier: Brandtech
110194 110294 110394 110494 110594 83330
75844-996PK 147.44 CAD
75844-996 75844-998 75845-000 75845-002 75845-004 75845-006
VITLAB® VITgrip™ Laboratory Bottles, Polypropylene, BrandTech
Bottles Plastic Bottles
VITgrip™ Polypropylene Laboratory Bottles feature an ergonomic grip, accurate graduations, and protection against liquid leakage.
  • Excellent handling
  • Leakproof sealing system
  • Easy readability
  • Robust and versatile
  • Tamper evident closure
  • Recyclable packaging
The slim waist of the VITgrip™ lab bottle greatly improves handling.
Manufactured from shatterproof polypropylene with good chemical resistance, VITgrip™ lab bottles are suitable for contact with foodstuffs (according to regulation EU No. 10/2011). The hydrophobic material and lack of dirt-More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable