Kimtech™ A7 Certified Liquid Barrier Gown, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Microflex
47992 47993 47994 47992 47993 47994
75870-726CS 812.26 CAD
75870-726 75870-728 75870-730 CA75870-726 CA75870-728 CA75870-730
Kimtech™ A7 Certified Liquid Barrier Gown, KIMBERLY-CLARK PROFESSIONAL®
Disposable Garments
When working with blood-borne pathogens, biologics, or other harmful liquids, workers must seek advanced protection against spills and splashes. The Kimtech™ A7 certified liquid barrier gown is an ideal solution.
  • Bulk packaged
  • Thumb loops to maintain glove and garment interface
  • Seamless front and closed back
  • Easy to don and doff design
Kimtech™ A7 certified liquid barrier gowns deliver stringent barrier protection for people and superior contamination control for any facility. This protective gown has plenty of innovative features, including aMore Product Information