SP Wilmad-LabGlass Reaction Kettle Heating Mantles, SP Industries
Supplier: SP Industries
LG-8883-110 LG-8883-108 LG-8883-106 LG-8883-100 LG-8883-102
14200-990EA 1425.22 CAD
14200-990 14200-550 14200-764 14200-714 14200-728
SP Wilmad-LabGlass Reaction Kettle Heating Mantles, SP Industries
Heaters Heating Mantles
Designed for use with cylindrical reaction kettles LG-8071 through LG-8082, these heating mantles have an aluminum housing exterior.
  • Includes detachable 4 foot, 3-wire cord and locking connector
They are equipped with a 3-wire terminal with one grounded to housing.